siRNA Details
| Gene Symbol: Zfp90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025765.1 | NP_001020936.1 | 5 | 1400 | ||
| XM_006255565.2 | XP_006255627.1 | 3 | 2883 | |||
| XM_006255566.2 | XP_006255628.1 | 4 | 1465 | |||
| GenBank mRNA | BC097467.1 | - | 5 | 1400 | 7457672 | |

Zfp90
Genomic Map