siRNA Details
| Gene Symbol: Tmem170a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134608.1 | NP_001128080.1 | 2 | 205 | ||
| XM_008772528.1 | XP_008770750.1 | 2 | 205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem170a| Gene Symbol: Tmem170a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134608.1 | NP_001128080.1 | 2 | 205 | ||
| XM_008772528.1 | XP_008770750.1 | 2 | 205 | |||