siRNA Details
| Gene Symbol: Fbxo31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044259.2 | NP_001037724.2 | 6 | 775 | ||
| XM_006255719.2 | XP_006255781.1 | 6 | 1769 | |||
| XM_006255720.2 | XP_006255782.1 | 5 | 1682 | |||
| GenBank mRNA | BC110046.1 | - | 4 | 881 | 7302542 | |
| BC166840.1 | - | 6 | 775 | 9027777 | ||

Fbxo31
Genomic Map