siRNA Details
| Gene Symbol: Spata2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109133.1 | NP_001102603.1 | 2 | 1531 | ||
| XM_006255781.2 | XP_006255843.1 | 3 | 1984 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata2L| Gene Symbol: Spata2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109133.1 | NP_001102603.1 | 2 | 1531 | ||
| XM_006255781.2 | XP_006255843.1 | 3 | 1984 | |||