siRNA Details
| Gene Symbol: RGD1559896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109134.1 | NP_001102604.1 | 1 | 245 | ||
| XM_008772642.1 | XP_008770864.1 | 1 | 280 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1559896| Gene Symbol: RGD1559896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109134.1 | NP_001102604.1 | 1 | 245 | ||
| XM_008772642.1 | XP_008770864.1 | 1 | 280 | |||