siRNA Details
| Gene Symbol: Cpne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256466.1 | NP_001243395.1 | 1, 2 | 178 | ||
| XM_008772319.1 | XP_008770541.1 | 2, 3 | 355 | |||
| GenBank mRNA | AM747280.1 | - | 1, 2 | 178 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpne2| Gene Symbol: Cpne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256466.1 | NP_001243395.1 | 1, 2 | 178 | ||
| XM_008772319.1 | XP_008770541.1 | 2, 3 | 355 | |||
| GenBank mRNA | AM747280.1 | - | 1, 2 | 178 | ||