siRNA Details
| Gene Symbol: Tmem200a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109135.1 | NP_001102605.1 | 2 | 2263 | ||
| XM_006227690.2 | XP_006227752.1 | 3 | 3641 | |||
| XM_008758623.1 | XP_008756845.1 | 2 | 3191 | |||
| XM_008758624.1 | XP_008756846.1 | 3 | 1938 | |||
| XM_008758625.1 | XP_008756847.1 | 4 | 3746 | |||

Tmem200a
Genomic Map