siRNA Details
| Gene Symbol: Ahrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024285.1 | NP_001019456.1 | 5 | 476 | ||
| GenBank mRNA | AB174900.2 | - | 5 | 476 | ||
| AY367561.1 | - | 5 | 476 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ahrr| Gene Symbol: Ahrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024285.1 | NP_001019456.1 | 5 | 476 | ||
| GenBank mRNA | AB174900.2 | - | 5 | 476 | ||
| AY367561.1 | - | 5 | 476 | |||