siRNA Details
| Gene Symbol: Zdhhc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039342.2 | NP_001034431.2 | 6 | 812 | ||
| XM_006227796.2 | XP_006227858.1 | 5 | 824 | |||
| XM_006227797.2 | XP_006227859.1 | 5 | 773 | |||
| GenBank mRNA | AY886527.1 | - | 6 | 844 | ||

Zdhhc11
Genomic Map