siRNA Details
| Gene Symbol: Zdhhc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039342.2 | NP_001034431.2 | 6, 7 | 841 | ||
| XM_006227796.2 | XP_006227858.1 | 5, 6 | 853 | |||
| XM_006227797.2 | XP_006227859.1 | 5, 6 | 802 | |||
| GenBank mRNA | AY886527.1 | - | 6, 7 | 873 | ||

Zdhhc11
Genomic Map