siRNA Details
| Gene Symbol: Zdhhc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039343.1 | NP_001034432.1 | 1 | 1015 | ||
| XM_006227884.2 | XP_006227946.1 | 1 | 978 | |||
| XM_006227885.2 | XP_006227947.1 | 1 | 983 | |||
| XM_006227886.2 | XP_006227948.1 | 1 | 982 | |||
| GenBank mRNA | AY886530.1 | - | 1 | 1015 | ||

Zdhhc14
Genomic Map