siRNA Details
| Gene Symbol: Sytl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127560.1 | NP_001121032.1 | 13 | 1673 | ||
| XM_006227888.2 | XP_006227950.1 | 14 | 2229 | |||
| XM_006227891.2 | XP_006227953.1 | 14 | 2226 | |||
| GenBank mRNA | BC166706.1 | - | 13 | 1673 | 5599231 | |

Sytl3
Genomic Map