siRNA Details
Gene Symbol: Ermard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728660.4 | XP_002728706.1 | 12 | 1244 | ||
XM_006227972.2 | XP_006228034.1 | 12 | 1244 | |||
XM_006227974.1 | XP_006228036.1 | 12 | 1349 | |||
XM_006227975.2 | XP_006228037.1 | 12 | 1186 | |||
XM_006227976.1 | XP_006228038.1 | 10 | 2479 | |||
XM_008758815.1 | XP_008757037.1 | 10 | 957 | |||
XM_008758816.1 | XP_008757038.1 | 8 | 791 | |||
XM_008758817.1 | XP_008757039.1 | 8 | 729 |
