siRNA Details
| Gene Symbol: Znf761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228009.2 | XP_006228071.1 | 4 | 1659 | ||
| XM_006228010.2 | XP_006228072.1 | 5 | 2583 | |||
| XM_006228011.2 | XP_006228073.1 | 5 | 2578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znf761| Gene Symbol: Znf761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228009.2 | XP_006228071.1 | 4 | 1659 | ||
| XM_006228010.2 | XP_006228072.1 | 5 | 2583 | |||
| XM_006228011.2 | XP_006228073.1 | 5 | 2578 | |||