siRNA Details
| Gene Symbol: Zfp61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017512.2 | NP_001017512.2 | 6 | 1911 | ||
| XM_008758962.1 | XP_008757184.1 | 7 | 2293 | |||
| GenBank mRNA | BC079015.1 | - | 6 | 1858 | 7113620 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp61| Gene Symbol: Zfp61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017512.2 | NP_001017512.2 | 6 | 1911 | ||
| XM_008758962.1 | XP_008757184.1 | 7 | 2293 | |||
| GenBank mRNA | BC079015.1 | - | 6 | 1858 | 7113620 | |