siRNA Details
Gene Symbol: LOC102553892 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759075.1 | XP_008757297.1 | 2 | 190 | ||
XM_008759076.1 | XP_008757298.1 | 2 | 190 |
Gene Symbol: RGD1564380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758911.1 | XP_008757133.1 | 3 | 515 | ||
XM_008758912.1 | XP_008757134.1 | 3 | 523 | |||
XM_008758913.1 | XP_008757135.1 | 3 | 376 | |||
XM_008758914.1 | XP_008757136.1 | 3 | 518 | |||
XM_008758915.1 | XP_008757137.1 | 3 | 515 |
Gene Symbol: RGD1565655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177686.1 | NP_001171157.2 | 3 | 469 | ||
XM_008758968.1 | XP_008757190.1 | 190 | ||||
GenBank mRNA | BC158626.1 | - | 3 | 469 | 7455240 |
