siRNA Details
| Gene Symbol: Dmrtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109140.1 | NP_001102610.1 | 7 | 923 | ||
| Gene Symbol: LOC100910371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244579.2 | XP_006244641.1 | 7 | 977 | ||
| XM_008766899.1 | XP_008765121.1 | 6 | 1576 | |||
| XM_008766900.1 | XP_008765122.1 | 6 | 1576 | |||

Dmrtc2
Genomic Map