siRNA Details
| Gene Symbol: Cldnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109143.1 | NP_001102613.1 | 2 | 258 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldnd2| Gene Symbol: Cldnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109143.1 | NP_001102613.1 | 2 | 258 | ||