siRNA Details
| Gene Symbol: Spib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024286.1 | NP_001019457.1 | 6 | 679 | ||
| XM_006229111.2 | XP_006229173.1 | 5 | 1428 | |||
| XM_006229112.2 | XP_006229174.1 | 6 | 748 | |||
| XM_006229113.2 | XP_006229175.1 | 5 | 1374 | |||
| XM_006229114.2 | XP_006229176.1 | 5 | 1373 | |||
| XM_006229115.2 | XP_006229177.1 | 6 | 693 | |||
| XM_008759404.1 | XP_008757626.1 | 5 | 1376 | |||
| XM_008759405.1 | XP_008757627.1 | 5 | 729 | |||
| GenBank mRNA | BC089881.1 | - | 6 | 679 | 7368473 | |

Spib
Genomic Map