siRNA Details
| Gene Symbol: Gas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127504.1 | NP_001120976.1 | 6 | 806 | ||
| XM_006229270.2 | XP_006229332.1 | 7 | 1271 | |||
| XM_006229271.2 | XP_006229333.1 | 6 | 725 | |||
| XM_006229273.2 | XP_006229335.1 | 7 | 1063 | |||
| XM_006229274.2 | XP_006229336.1 | 7 | 1590 | |||
| XM_008759407.1 | XP_008757629.1 | 6 | 804 | |||
| XM_008759408.1 | XP_008757630.1 | 6 | 725 | |||

Gas2
Genomic Map