siRNA Details
| Gene Symbol: Fam189a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759569.1 | XP_008757791.1 | 1, 2 | 264 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam189a1| Gene Symbol: Fam189a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759569.1 | XP_008757791.1 | 1, 2 | 264 | ||