siRNA Details
| Gene Symbol: Cers3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127561.1 | NP_001121033.1 | 72 | |||
| GenBank mRNA | BC160913.1 | - | 72 | 7457160 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cers3| Gene Symbol: Cers3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127561.1 | NP_001121033.1 | 72 | |||
| GenBank mRNA | BC160913.1 | - | 72 | 7457160 | ||