siRNA Details
| Gene Symbol: Acer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759809.1 | XP_008758031.1 | 8 | 627 | ||
| GenBank mRNA | FQ230480.1 | - | 6 | 474 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acer3| Gene Symbol: Acer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759809.1 | XP_008758031.1 | 8 | 627 | ||
| GenBank mRNA | FQ230480.1 | - | 6 | 474 | ||