siRNA Details
| Gene Symbol: RGD1564933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759847.1 | XP_008758069.1 | 1 | 117 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564933| Gene Symbol: RGD1564933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759847.1 | XP_008758069.1 | 1 | 117 | ||