siRNA Details
| Gene Symbol: LOC499240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024287.1 | NP_001019458.1 | 2, 3 | 236 | ||
| XM_008759786.1 | XP_008758008.1 | 2, 3 | 521 | |||
| GenBank mRNA | BC087685.1 | - | 2, 3 | 236 | 7309316 | |

LOC499240
Genomic Map