siRNA Details
| Gene Symbol: RGD1563217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109155.1 | NP_001102625.1 | 3 | 259 | ||
| XM_006230320.2 | XP_006230382.1 | 3 | 437 | |||
| XM_006230321.2 | XP_006230383.1 | 3 | 216 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1563217| Gene Symbol: RGD1563217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109155.1 | NP_001102625.1 | 3 | 259 | ||
| XM_006230320.2 | XP_006230382.1 | 3 | 437 | |||
| XM_006230321.2 | XP_006230383.1 | 3 | 216 | |||