siRNA Details
| Gene Symbol: RGD1560958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109157.1 | NP_001102627.1 | 4 | 388 | ||
| XM_006230418.2 | XP_006230480.1 | 3 | 466 | |||
| XM_008759915.1 | XP_008758137.1 | 3 | 394 | |||
| XM_008759916.1 | XP_008758138.1 | 3 | 885 | |||
| GenBank mRNA | BC091569.1 | - | 3 | 275 | ||

RGD1560958
Genomic Map