siRNA Details
| Gene Symbol: Fam24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109158.1 | NP_001102628.1 | 2 | 163 | ||
| XM_006230419.2 | XP_006230481.1 | 4 | 530 | |||
| XM_008759917.1 | XP_008758139.1 | 2 | 82 | |||
| GenBank mRNA | BC161873.1 | - | 4 | 471 | 7113177 | |
| BC166439.1 | - | 2 | 90 | 7453416 | ||

Fam24a
Genomic Map