siRNA Details
| Gene Symbol: Mapk1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122782.1 | NP_001116254.1 | 1 | 729 | ||
| XM_006230487.2 | XP_006230549.1 | 3 | 1873 | |||
| XM_006230488.2 | XP_006230550.1 | 2 | 2164 | |||
| XM_006230489.2 | XP_006230551.1 | 4 | 1038 | |||
| XM_006230490.2 | XP_006230552.1 | 3 | 915 | |||
| XM_006230491.2 | XP_006230553.1 | 2 | 794 | |||
| XM_008760006.1 | XP_008758228.1 | 3 | 1329 | |||
| XM_008760007.1 | XP_008758229.1 | 2 | 1206 | |||
| GenBank mRNA | BC158819.1 | - | 1 | 729 | 7374980 | |

Mapk1ip1
Genomic Map