siRNA Details
| Gene Symbol: Lrrc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113789.1 | NP_001107261.1 | 2 | 341 | ||
| NM_001143756.1 | NP_001137228.1 | 2 | 180 | |||
| XM_006230495.2 | XP_006230557.1 | 2 | 254 | |||
| XM_008760008.1 | XP_008758230.1 | 2 | 1082 | |||
| XM_008760009.1 | XP_008758231.1 | 2 | 221 | |||
| GenBank mRNA | BC158629.1 | - | 1 | 78 | 7456037 | |
| CK469366.1 | - | 2 | 180 | |||

Lrrc27
Genomic Map