siRNA Details
| Gene Symbol: Mob2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109159.1 | NP_001102629.1 | 364 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mob2| Gene Symbol: Mob2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109159.1 | NP_001102629.1 | 364 | |||