siRNA Details
| Gene Symbol: Tut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033901.1 | NP_001029073.1 | 2 | 233 | ||
| GenBank mRNA | BC104695.1 | - | 2 | 233 | 7111833 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tut1| Gene Symbol: Tut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033901.1 | NP_001029073.1 | 2 | 233 | ||
| GenBank mRNA | BC104695.1 | - | 2 | 233 | 7111833 | |