siRNA Details
| Gene Symbol: LOC100910973 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FM045317.1 | - | 2 | 77 | ||
| FM097063.1 | - | 2 | 95 | |||
| FM098643.1 | - | 2 | 267 | |||
| FM110991.1 | - | 2 | 70 | |||
| FQ212263.1 | - | 2 | 91 | |||
| FQ230677.1 | - | 2 | 91 | |||
| FQ231880.1 | - | 2 | 89 | |||
| FQ234947.1 | - | 2 | 87 | |||

LOC100910973
Genomic Map