siRNA Details
| Gene Symbol: Ms4a13-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231104.2 | XP_006231166.1 | 3 | 368 | ||
| XM_006231105.2 | XP_006231167.1 | 3 | 428 | |||
| XM_008760270.1 | XP_008758492.1 | 3 | 428 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a13-ps1| Gene Symbol: Ms4a13-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231104.2 | XP_006231166.1 | 3 | 368 | ||
| XM_006231105.2 | XP_006231167.1 | 3 | 428 | |||
| XM_008760270.1 | XP_008758492.1 | 3 | 428 | |||