siRNA Details
| Gene Symbol: Fam111a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109163.1 | NP_001102633.1 | 3 | 1820 | ||
| XM_006231125.1 | XP_006231187.1 | 3 | 1850 | |||
| GenBank mRNA | FQ230484.1 | - | 3 | 1818 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam111a| Gene Symbol: Fam111a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109163.1 | NP_001102633.1 | 3 | 1820 | ||
| XM_006231125.1 | XP_006231187.1 | 3 | 1850 | |||
| GenBank mRNA | FQ230484.1 | - | 3 | 1818 | ||