siRNA Details
| Gene Symbol: Fam111a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109163.1 | NP_001102633.1 | 2 | 206 | ||
| XM_006231125.1 | XP_006231187.1 | 2 | 236 | |||
| GenBank mRNA | FQ226103.1 | - | 2 | 205 | ||
| FQ230484.1 | - | 2 | 204 | |||

Fam111a
Genomic Map