siRNA Details
| Gene Symbol: Blnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025767.1 | NP_001020938.1 | 4, 5 | 301 | ||
| GenBank mRNA | BC098790.1 | - | 4, 5 | 301 | 7375523 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Blnk| Gene Symbol: Blnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025767.1 | NP_001020938.1 | 4, 5 | 301 | ||
| GenBank mRNA | BC098790.1 | - | 4, 5 | 301 | 7375523 | |