siRNA Details
| Gene Symbol: Fam178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134612.1 | NP_001128084.1 | 12 | 2793 | ||
| XM_006231550.2 | XP_006231612.1 | 11 | 2785 | |||
| XM_006231551.2 | XP_006231613.1 | 11 | 2781 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam178a| Gene Symbol: Fam178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134612.1 | NP_001128084.1 | 12 | 2793 | ||
| XM_006231550.2 | XP_006231612.1 | 11 | 2785 | |||
| XM_006231551.2 | XP_006231613.1 | 11 | 2781 | |||