siRNA Details
| Gene Symbol: Hsf2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127683.1 | NP_001121155.1 | 5 | 534 | ||
| XM_008772826.1 | XP_008771048.1 | 6 | 770 | |||
| GenBank mRNA | BC158765.1 | - | 5 | 521 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsf2bp| Gene Symbol: Hsf2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127683.1 | NP_001121155.1 | 5 | 534 | ||
| XM_008772826.1 | XP_008771048.1 | 6 | 770 | |||
| GenBank mRNA | BC158765.1 | - | 5 | 521 | ||