siRNA Details
Gene Symbol: Sumo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024295.1 | NP_001019466.1 | 1, 2 | 103 | ||
GenBank mRNA | BC083728.1 | - | 1, 2 | 103 | 7191303 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sumo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024295.1 | NP_001019466.1 | 1, 2 | 103 | ||
GenBank mRNA | BC083728.1 | - | 1, 2 | 103 | 7191303 |