siRNA Details
| Gene Symbol: Spatc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024296.1 | NP_001019467.1 | 2 | 565 | ||
| GenBank mRNA | BC079291.1 | - | 2 | 565 | 7134390 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spatc1l| Gene Symbol: Spatc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024296.1 | NP_001019467.1 | 2 | 565 | ||
| GenBank mRNA | BC079291.1 | - | 2 | 565 | 7134390 | |