siRNA Details
| Gene Symbol: RGD1564149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079708.1 | NP_001073176.1 | 1 | 227 | ||
| GenBank mRNA | BC128753.1 | - | 1 | 227 | 7100076 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564149| Gene Symbol: RGD1564149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079708.1 | NP_001073176.1 | 1 | 227 | ||
| GenBank mRNA | BC128753.1 | - | 1 | 227 | 7100076 | |