siRNA Details
| Gene Symbol: Prmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025144.1 | NP_001020315.1 | 9 | 1112 | ||
| GenBank mRNA | BC079112.1 | - | 9 | 1112 | 7125748 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prmt2| Gene Symbol: Prmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025144.1 | NP_001020315.1 | 9 | 1112 | ||
| GenBank mRNA | BC079112.1 | - | 9 | 1112 | 7125748 | |