siRNA Details
| Gene Symbol: Gstt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137643.1 | NP_001131115.1 | 1, 2 | 564 | ||
| GenBank mRNA | FQ225600.1 | - | 1, 2 | 227 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gstt3| Gene Symbol: Gstt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137643.1 | NP_001131115.1 | 1, 2 | 564 | ||
| GenBank mRNA | FQ225600.1 | - | 1, 2 | 227 | ||