siRNA Details
| Gene Symbol: Lims1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145456.1 | NP_001138928.1 | 3 | 344 | ||
| XM_006256385.2 | XP_006256447.1 | 3 | 2246 | |||
| XM_006256386.2 | XP_006256448.1 | 3 | 2246 | |||
| XM_006256387.2 | XP_006256449.1 | 3 | 2243 | |||
| XM_006256388.2 | XP_006256450.1 | 3 | 2110 | |||
| XM_006256389.2 | XP_006256451.1 | 3 | 337 | |||
| XM_006256390.2 | XP_006256452.1 | 3 | 1917 | |||
| XM_006256391.2 | XP_006256453.1 | 3 | 1916 | |||
| GenBank mRNA | FJ640984.1 | - | 3 | 344 | ||
| FJ640985.1 | - | 3 | 423 | |||
| FQ222665.1 | - | 3 | 432 | |||

Lims1
Genomic Map