siRNA Details
| Gene Symbol: Mettl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025038.1 | NP_001020209.1 | 2, 3 | 418 | ||
| GenBank mRNA | BC091261.1 | - | 2, 3 | 418 | 7377954 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl24| Gene Symbol: Mettl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025038.1 | NP_001020209.1 | 2, 3 | 418 | ||
| GenBank mRNA | BC091261.1 | - | 2, 3 | 418 | 7377954 | |