siRNA Details
Gene Symbol: Zfyve16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191958.1 | NP_001178887.1 | 3 | 1695 | ||
XM_006231783.2 | XP_006231845.1 | 2 | 1605 | |||
XM_006231784.2 | XP_006231846.1 | 4 | 1778 | |||
XM_006231785.2 | XP_006231847.1 | 3 | 1664 | |||
XM_008760640.1 | XP_008758862.1 | 3 | 2042 |
