siRNA Details
| Gene Symbol: Zfyve16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191958.1 | NP_001178887.1 | 9 | 3524 | ||
| XM_006231783.2 | XP_006231845.1 | 8 | 3434 | |||
| XM_006231784.2 | XP_006231846.1 | 10 | 3607 | |||
| XM_006231785.2 | XP_006231847.1 | 9 | 3493 | |||
| XM_008760640.1 | XP_008758862.1 | 9 | 3871 | |||

Zfyve16
Genomic Map