siRNA Details
| Gene Symbol: Zfyve16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191958.1 | NP_001178887.1 | 3 | 665 | ||
| XM_006231783.2 | XP_006231845.1 | 2 | 575 | |||
| XM_006231784.2 | XP_006231846.1 | 4 | 748 | |||
| XM_006231785.2 | XP_006231847.1 | 3 | 634 | |||
| XM_008760640.1 | XP_008758862.1 | 3 | 1012 | |||

Zfyve16
Genomic Map