siRNA Details
| Gene Symbol: Spz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024297.1 | NP_001019468.1 | 1 | 215 | ||
| GenBank mRNA | BC079262.1 | - | 1 | 215 | 7133509 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spz1| Gene Symbol: Spz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024297.1 | NP_001019468.1 | 1 | 215 | ||
| GenBank mRNA | BC079262.1 | - | 1 | 215 | 7133509 | |